Learn More
Medchemexpress LLC Indole-3-acetylglycine | 13113-08-1 | MFCD00079392 | 95.0% | 232.24 g/mol | C12H12N2O3 | 1 G

Supplier: Medchemexpress LLC HYW3174141G
Indole-3-acetylglycine is an indole-containing N-acyl amino acid used as a biochemical assay reagent in research. Supplied as a solid powder, it is used as an analytical standard or reagent in metabolic and urine studies, and is intended for research use only.
- Indole-derived N-acyl amino acid suitable for biochemical assays.
- Used as an analytical standard or reagent in metabolic and urine studies.
- Supplied as a 1 g powder with typical purity around 95%.
- Molecular formula C12H12N2O3 and molar mass ≈232.24 g/mol.
- CAS number 13113-08-1 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.