Learn More
Medchemexpress LLC Indole-3-acetylglycine | 13113-08-1 | MFCD00079392 | 95.0% | 232.24 g/mol | C12H12N2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW31741450MG
Encompass_Preferred
Indole-3-acetylglycine is an indole-derived biochemical assay reagent used as a reference standard and metabolite in analytical and biochemical research. It has the molecular formula C12H12N2O3 and a molecular weight of 232.24 g/mol. Typical uses include metabolite profiling, analytical method development, and as a standard in HPLC and mass spectrometry assays.
  • High chemical purity suitable for analytical applications.
  • Well-characterized physical properties for reliable identification.
  • Available in multiple small pack sizes for flexible laboratory use.
  • Compatible with common analytical techniques such as HPLC and MS.
  • Stable when stored under recommended conditions for reproducible results.
Catalog No. 50-002-80662