Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-3-acetylglycine | 13113-08-1 | MFCD00079392 | 95.0% | 232.24 g/mol | C12H12N2O3 | 500 MG
SDP

Supplier:  Medchemexpress LLC HYW317414500MG

Encompass_Preferred

Indole-3-acetylglycine is a small-molecule biochemical assay reagent used in analytical workflows and research applications. It is supplied with supporting analytical documentation and multiple package sizes to accommodate assay development and mid-scale laboratory use.

  • Biochemical assay reagent for research use.
  • Molecular formula C12H12N2O3 and molecular weight 232.24 g/mol.
  • CAS number 13113-08-1 for unambiguous identification.
  • Typical purity 95.0% as listed by the manufacturer.
  • Available in multiple sizes including 500 MG for mid-scale experiments.
  • Supporting documentation available: COA, SDS, LC-MS, and HNMR.

Catalog No. 50-002-81867


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.