Learn More
Medchemexpress LLC Indole-3-amidoxime | 95649-37-9 | MFCD00460181 | 96.3% | 175.19 g/mol | C9H9N3O | 100 MG

Supplier: Medchemexpress LLC HYW590911100MG
Indole-3-amidoxime is a synthetic intermediate used in pharmaceutical and medicinal chemistry to build and modify small-molecule drug candidates. It is supplied for research use and not intended for human therapeutic applications.
- Used as a building block for pharmaceutical synthesis and medicinal chemistry research.
- Molecular formula C9H9N3O; molecular weight 175.19 g/mol.
- CAS number 95649-37-9 for unambiguous identification.
- Available in multiple pack sizes, including 100 MG for small-scale experiments.
- Typical purity approximately 96.3% as supplied.
- Sparingly soluble in aqueous media; better solubility in DMSO and DMF.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.