Learn More
Bioworld Indole-3-Butyric Acid (IBA), 25 g
Supplier:  Bioworld 306310112
Encompass_Preferred
Indole-3-butyric acid (IBA)Synonym: 4-(3-Indolyl)butanoic acidSynthetic AuxinPlant tissue culture testedAppearance: white to light tan powderMelting Point: 124.0-125.5 °CSoluble in ethanol or 1N NaOH
Catalog No. 50-255-051