Learn More
Medchemexpress LLC Indole-3-carboxaldehyde-13C8 | MFCD00005622 | 98.0% | 145.16 | C9H7NO (13C8) | 500ug
SDP
Supplier:  Medchemexpress LLC HYW750674500UG
Encompass_Preferred
Indole-3-carboxaldehyde-13C8 (3-Formylindole-13C8) is the 13C-labeled Indole-3-carboxaldehyde (HY-W007376) Indole-3-carboxaldehyde a banlangen extract is the product of the oxidative degradation of indole-3-acetic acid (IAA) by crude enzyme preparations from etiolated pea seedlings Indole-3-carboxaldehyde is a biochemical used to prepare analogs of the indole phytoalexin cyclobrassinin Indole-3-carboxaldehyde also enhances the epithelial barrier and anti-inflammatory activity in the intestinal tract[1][2]
Catalog No. 50-002-69042