Learn More
Cayman Chemical Indole-3-carboxaldehyde 25g
SDP
Supplier:  Cayman Chemical 3132825 G
Encompass_Preferred
An active tryptophan metabolite and synthetic intermediate; increases colonic production of IL-22 and restores colonization resistance to C. albicans infection in a wild-type, but not Ahr-/-, mouse model of mucosal candidiasis at 18 mg/kg; reduces intestinal mucosal damage in a mouse model of DSS-induced colitis
Catalog No. 50-299-1121