Learn More
Apexbio Technology LLC Indole-3-carboxaldehyde 487-89-8 100mg
SDP
Supplier:  Apexbio Technology LLC C6813100
Encompass_Preferred
Indole-3-carboxaldehyde (CAS 487-89-8) also known as 3-formylindole is an indole derivative produced during the oxidative metabolism of indole-3-acetic acid in plants This compound modulates intestinal epithelial barrier function and exhibits anti-inflammatory properties as demonstrated in cellular and animal studies Indole-3-carboxaldehyde is commonly employed in biomedical research to investigate mechanisms underlying gut barrier protection the regulation of mucosal immunity and the development of indole-based analogs for plant and microbial defense studies
Catalog No. 50-000-89816