Learn More
Medchemexpress LLC Indole-3-lactic acid | 1821-52-9 | MFCD00005642 | 100.0% | 205.21 g/mol | C11H11NO3 | 5 MG

Supplier: Medchemexpress LLC HY1130995MG
Indole-3-lactic acid is a tryptophan catabolite used as an analytical and research standard. Supplied as a high-purity off-white to light-yellow solid, it is employed in biochemical and metabolite studies and formulation research. Reported biological effects include anti-inflammatory activity and potential antiviral properties.
- High purity (99.95%) analytical standard.
- Molecular weight 205.21 g/mol.
- Chemical formula C11H11NO3.
- Off-white to light-yellow solid appearance.
- Available in small package sizes suitable for analytical work.
- Soluble in DMSO at 100 mg/mL; sonication may be required.
- Powder storage: 4°C for 2 years, -20°C for 3 years; in solution storage: -80°C for 1 year.
- Typical applications: metabolite reference, analytical assays, and biochemical research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.