Learn More
Medchemexpress LLC Indole-3-methanamine | 22259-53-6 | 99.1% | 146.19 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY33893
Encompass_Preferred
Indole-3-methanamine is identified as a potential biomarker for the consumption of foods such as barley, cereals, and cereal products. This product is intended for research use only.
  • Potential biomarker for food consumption.
  • Store at -20°C, protected from light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Ships at room temperature in continental US.
Catalog No. 50-004-02689