Learn More
Medchemexpress LLC Indole-3-methanamine | 22259-53-6 | 99.1% | 146.19 | 500 MG

Supplier: Medchemexpress LLC HY33893500MG
Indole-3-methanamine is a potential biomarker for the consumption of foods such as barley, cereals, and cereal products.
- Target: Endogenous metabolite
- Research areas: Metabolic disease
- Category: Natural products
- Source classification: Endogenous metabolite
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.