Learn More
Medchemexpress LLC Indole-3-methanamine | 22259-53-6 | 99.14% | 146.19 | 25 G

Supplier: Medchemexpress LLC 25GHY33893
Indole-3-methanamine is an endogenous metabolite and a potential biomarker for the consumption of foods such as barley, cereals, and cereal products.
- Potential biomarker for food consumption
- For research use only
- Ships at room temperature in continental US
- Storage at -20°C, protected from light
- Soluble in DMSO: 50 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.