Learn More
Medchemexpress LLC Indole-3-propionic acid | 830-96-6 | MFCD00005660 | 99.8% | 189.21 g/mol | C11H11NO2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW015229R
Encompass_Preferred
3-Indolepropionic acid (indole-3-propionic acid, IPA) is an indole-derived small molecule studied for its antioxidant and neuroprotective properties. It is used in biochemical and pharmacological research as a radical scavenger and as a metabolite/biomarker in metabolic and neurodegenerative disease studies.
  • High purity suitable for analytical and biological assays.
  • Potent antioxidant and hydroxyl radical scavenger.
  • Useful in neuroprotection and Alzheimer's disease research.
  • Available as solid and as DMSO solution for flexible handling.
  • Molecular formula C11H11NO2; molecular weight 189.21 g/mol.
Catalog No. 50-003-02346