Learn More
Medchemexpress LLC Indole-d7 | 73509-20-3 | 124.19 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW001132S45MG
Encompass_Preferred
Indole-d7 is the deuterium labeled Indole, an endogenous metabolite. It serves as a valuable tool for quantitative analysis in various spectroscopic methods, including NMR, GC-MS, and LC-MS. The deuteration of Indole-d7 holds significance due to its potential influence on the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Endogenous metabolite
  • Suitable for tracer applications
  • Internal standard for quantitative analysis
  • Used in NMR, GC-MS, and LC-MS
  • Potential for pharmacokinetic and metabolic studies
Catalog No. 50-004-47567