Learn More
Medchemexpress LLC Indoleamine 2,3-dioxygenase antibody (YA3100)(PBS only) | 50 UL

Supplier: Medchemexpress LLC HYP8335550UL
Indoleamine 2,3-dioxygenase antibody (YA3100) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting indoleamine 2,3-dioxygenase. Supplied in PBS, pH 7.4, this antibody is crucial in the study of tryptophan catabolism. The enzyme modulates T-cell behavior and is involved in various pathophysiological processes including antimicrobial and antitumor defense, neuropathology, immunoregulation, and antioxidant activity.
- Host: Rabbit
- Reactivity: Human
- Conjugation: Non-conjugated
- SwissProt ID: P14902
- Research field: Signal transduction
- Appearance: Liquid
- Storage: Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles.
- Shipping: With blue ice.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.