Learn More
Cayman Chemical IndolImIn-214 10mg
SDP
Supplier:  Cayman Chemical 3795310 MG
Encompass_Preferred
A genotoxic gut microbiota metabolite; increases γH2AX levels in HeLa cells at 25 and 100 UG/ml; increases the tail DNA percentage in a comet assay using HeLa cells at 100 UG/ml
Catalog No. 50-291-8085