Learn More
Selleck Chemical LLC Indomethacin S1723-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S172310MM/1ML
Encompass_Preferred
Indomethacin is a nonselective COX1 and COX2 inhibitor with IC50 of 0 1 g/mL and 5 g/mL respectively used to reduce fever pain stiffness and swelling
Catalog No. 50-311-0656