Learn More
Sigma Aldrich Fine Chemicals Biosciences Indomethacin 98.5-100.5 in25G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences I737825G
Encompass
Indomethacin is an inhibitor of the enzyme cyclooxygenase 1 and 2. Indomethacin elicits side effects in gastrointestinal tract kidney and cerebrum. Indomethacin along with ibuprofen is effective for treating patent ductus arteriosus (PDA) in infants with respiratory distress syndrome. Rectal indomethacin is effective in treating endoscopic retrograde cholangiography (ERCP) induced pancreatitis.
Catalog No. 50-290-2271