Learn More
Medchemexpress LLC Indomethacin-d4 methyl ester | 1217064-61-3 | 98.4% | 375.84 g/mol | C20H14D4ClNO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14397S15MG
Encompass_Preferred
Indomethacin-d4 methyl ester is a deuterium-labeled analogue of indomethacin supplied as a stable-isotope reference standard for analytical applications. It is provided as a white to off-white solid and intended for use as an internal standard in LC-MS/MS and other mass spectrometry assays requiring accurate quantification.
  • Deuterium-labeled indomethacin methyl ester for isotope-dilution assays.
  • Suitable as an internal standard for LC-MS and GC-MS quantification.
  • High purity (98.4%).
  • Molecular formula C20H14D4ClNO4; molecular weight 375.84 g/mol.
  • Supplied as a white to off-white solid in 5 MG packaging.
Catalog No. 50-002-21047