Learn More
Cayman Chemical IndomethacIn heptyl ester 10mg
SDP
Supplier:  Cayman Chemical 7027110 MG
Encompass_Preferred
A potent, non-selective COX-2 inhibitor (IC50 = 0.04 µM); >1,700 times more potent as an inhibitor of COX-2 than COX-1
Catalog No. 50-289-2858