Learn More
Medchemexpress LLC Indomethacin (Standard) | 53-86-1 | 99.8% | C₁₉H₁₆ClNO₄ | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14397R100MG
Encompass_Preferred
Indomethacin (Standard) is the analytical standard for Indomethacin, intended for research and analytical applications. It is a potent, orally active COX1/2 inhibitor with IC50 values of 18 nM for COX-1 and 26 nM for COX-2. This compound exhibits anticancer and anti-infective activities and is suitable for research in cancer, inflammation, and viral infection.
  • Analytical standard for Indomethacin.
  • Potent, orally active COX1/2 inhibitor.
  • Exhibits anticancer activity.
  • Shows anti-infective activity.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-55042