Learn More
Medchemexpress LLC Indotecan (hydrochloride) | 1228035-68-4 | 99.8% | 514.95 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY18351A
Encompass_Preferred
Indotecan hydrochloride, an indenoisoquinoline derivative, is a potent Topoisomerase I inhibitor. It has IC50s of 300, 1200, and 560 nM for P388, HCT116, and MCF-7 cell lines, respectively. This compound prevents the relaxation of supercoiled DNA and can be utilized for research on visceral leishmaniasis.
  • Potent Topoisomerase I inhibitor.
  • Effective in preventing relaxation of supercoiled DNA.
  • Can be used for research on visceral leishmaniasis.
Catalog No. 50-003-85927