Learn More
Medchemexpress LLC Indotecan hydrochloride | 1228035-68-4 | MFCD25977124 | 99.8% | 514.95 | C26H27ClN2O7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY18351A10MG
Encompass_Preferred
Indotecan hydrochloride is an indenoisoquinoline-derived topoisomerase I inhibitor supplied as the hydrochloride salt for research use. It is intended for preclinical and biochemical studies of topoisomerase I activity, DNA damage responses, and related pharmacology, and is provided in small-mass vials suitable for laboratory experiments.
  • High purity (99.8%), suitable for research applications.
  • Provided as the hydrochloride salt for improved handling and solubility.
  • Potent topoisomerase I inhibitor for preclinical DNA damage studies.
  • Characterized by CAS number 1228035-68-4, formula C26H27ClN2O7, and molecular weight 514.95.
  • Supplied in small, convenient vial sizes for laboratory use.
Catalog No. 50-002-95271