Learn More
Hach Company Induct Electrodeless Sens-San
Supplier:  Hach Company 3705E2T20P
Encompass
Sanitary Inductive Electrodeless Conductivity Finished Sensor-PPPolypropylene Sanitary (CIP) Flange Mounting Style 6m (20 ft) Analog Cable Paper Tag Electrodeless Conductivity Sensor
Catalog No. 50-002-07991