Learn More
Uline Induction Seal Caps | White | 150
Supplier:  Uline S25583CS
Encompass
White polypropylene caps with continuous thread ribbed sides and foil liners, FDA compliant, Use with Handheld Induction Sealer to create an airtight tamper-evident seal
Catalog No. NC2954468