Learn More
MSC Nilfisk Critical Area Vacuum, 9 gal, 1 1/2 hp, HEPA, Explosion-Proof
Supplier:  MSC 35702190
Encompass
This critical area vacuum is a dry pickup vacuum designed for hazardous location safe applications. It features a stainless steel tank with a 9-gallon dry capacity and HEPA filtration, ensuring efficient and safe debris collection. Powered by a 1 1/2 hp motor, it operates on 120V AC and provides 103 cfm airflow and 65 inches of static pressure.
  • Dry pickup type
  • Stainless steel tank
  • 9 gal dry capacity
  • HEPA filtration for critical areas
  • 1 1/2 hp motor
  • 120V operation
  • Explosion-proof application
  • PullClean filter cleaning system
  • Fully bonded and grounded construction
  • Includes tool and accessory tray
  • 30 ft power cord
  • 10 ft hose with 1 1/2 in diameter
Catalog No. 50-283-8827
Add to Cart
Request a Quote