Learn More
GL Sciences Inc Inertsil ODS-4 Analytical Column, 2um, 2.1mm ID, 100mm Length
Supplier:  GL Sciences Inc 502081204
Encompass
nertsil ODS-4 delivers the same extreme inertness to any type of compounds just like InertSustain C18 along with unprecedented stability under 100 % aqueous mobile phases for qualitative and quantitative analysis. However, as the base silica gel and carbon loading are different on Inertsil ODS-4, differences in selectivity can be observed for certain analytes.
Catalog No. NC1820184