Learn More
Medchemexpress LLC HY-13311A 5mg Medchemexpress, Infigratinib phosphate CAS:1310746-10-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13311A5MG
Encompass_Preferred
Medchemexpress, HY-13311A 5mg Infigratinib phosphate CAS:1310746-10-1 Infigratinib phosphate (BGJ-398 phosphate) is a potent inhibitor of the FGFR family with IC 50 of 0.9 nM, 1.4 nM, 1 nM, and 60 nM for FGFR1 , FGFR2 , FGFR3 , and FGFR4 , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2662