Learn More
Electron Microscopy Sciences INFINITY2 5.0 Megapixel USB 2.0 Microscopy Camera CCD, Color
SDP
Supplier:  Electron Microscopy Sciences 95118
Encompass
Equipped with a high-quality, Sony CCD sensor, INFINITY2 CCD USB 2.0 cameras offer excellent sensitivity, high dynamic range, and a 12-bit digital output. Features include binning, auto white balance, full exposure control, programmable gain, sub-windowing and region of interest. Ideal camera for brightfield, darkfield, and DIC imaging applications.
Sensor: CCD
Type: Color
Catalog No. 50-190-2311