Learn More
Raybiotech Inc INFLAMMATION ARRAY C3 (4)
Supplier:  Raybiotech Inc AAHINF34
Encompass_Preferred
RayBio C-Series Human Inflammation Antibody Array 3 Kit. Detects 40 Human Inflammatory Factors. Suitable for all liquid sample types.
Catalog No. 50-104-3153