Learn More
Advanced ImmunoChemical, Inc. Influenza A (H1N1) Virus (Strain A/Beijing/262/95), 0.1mg
Supplier:  Advanced ImmunoChemical, Inc. 7IVAH1N120.1MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Antigens/ Infectious Diseases/ Influenza A, (H1N1) virus (strain A/Beijing/262/95). Source:Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza A virus, strain A/Beijing/262/95.Applications:Serological studies of influenza A virus, immunogen for antibody production.Analysis:Purity > 90 %Immunological activity:Tested with anti-influenza A monoclonal antibodies ( Cat. # 5-IVAN) in ELISA.Purification:Ultracentrifugation with 10 – 40 % sucrose gradient.Presentation:Presentation: STE, 0.09 % sodium azide (NaN3) and 0.005 % thimerosal.Inactivation:Thimerosal and beta propiolactone treatment. This product has been treated in a manner consistent with methods of inactivation. Generally accepted good laboratory practices appropriate to microbiological/viral safe handling practices and techniques are required at work.Storage Conditions:-20 °C (between -15 and -30 °C allowed).
Catalog No. 50-153-9829