Learn More
ACROBiosystems Influenza A [Darwin/6/2021] Neuraminidase (NA) Protein, His Tag, 100 μg
Supplier: ACROBiosystems NEEV5247100UG
Recombinant Influenza A (Darwin/6/2021) neuraminidase (NA) protein expressed in HEK293 cells with an N-terminal polyhistidine tag. Supplied lyophilized (100 μg), SPR-verified for sialic acid binding, and quality-tested for purity and endotoxin for use in binding assays and antibody characterization.
- Expressed in human 293 (HEK293) cells.
- Contains residues Thr 34-Ile 469 (GISAID accession EPI_ISL_3534319).
- N-terminal polyhistidine (His) tag for detection and purification.
- Calculated molecular weight 56.5 kDa; apparent 60-80 kDa on SDS-PAGE due to glycosylation.
- Purity greater than 90% by SDS-PAGE.
- Endotoxin less than 1.0 EU/μg (LAL / rFC).
- Lyophilized formulation with trehalose; store at -20°C or lower.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.