Learn More
Creative Biomart Recombinant Influenza A H7N7 (Netherlands) Antigen
Supplier:  Creative Biomart VAC0014
Encompass_Preferred
Recombinant Full-Length H7N7 A/Netherlands/219/03 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells and its Mw is approximately 70,000 Dalton.
Catalog No. 50-208-4854
 Restricted Item