Learn More
ATCC Influenza A virus; A/California/07/2009 pdm09 (H1N1)
Supplier:  ATCC VR1894
Encompass
Influenza A virus (H1N1) strain A/California/07/2009 (H1N1)pdm09 is propagated in SPF chicken embryonated eggs. This strain was isolated in 2009 from a human in California and was deposited by the Centers for Disease Control and Prevention. A/California/07/2009 (H1N1)pdm09 was recommended by the The US Food and Drug Administration for use in vaccines during the 2009-2010 and 2010-2011 influenza seasons. Use this Influenza A virus in your infectious and respiratory disease research and vaccine development.
Catalog No. 50-238-1745
 Restricted Item