Learn More
ATCC Influenza A virus (H1N1); A/Denver/1/57
Supplier:  ATCC VR546
Encompass
Influenza A virus (H1N1) strain A/Denver/1/57 is propagated in SPF embryonated chicken eggs. This strain was isolated from a human in February 1957 during an epidemic in Colorado, United States. This product has applications in influenza research.
Catalog No. 50-238-3596