Learn More
ATCC Influenza A virus (H1N1); A/Virginia/ATCC3/2009; RNA
Supplier:  ATCC VR1738D
Encompass
Genomic RNA isolated from a preparation of cell lysate and supernatant from MDCK cells infected with Influenza A virus (H1N1) strain A/Virginia/ATCC3/2009 (ATCC VR-1738). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-1967