Learn More
ATCC Influenza A virus (H1N1); A/WS/33
Supplier:  ATCC VR825
Encompass
Influenza A virus (H1N1) strain A/WS/33 is propagated in SPF chicken embryonated eggs. This strain was isolated in 1933 from the throat washings of a human patient with influenza. This product is whole-genome sequenced and has applications in influenza research.
Catalog No. 50-238-4039