Learn More
ATCC Influenza A virus (H3N2); A/Hong Kong/8/68; RNA
Supplier:  ATCC VR1679D
Encompass
Genomic RNA isolated from a preparation of cell lysate and supernatant from MDCK cells (ATCC CCL-34) infected with Influenza A virus A/Hong Kong/8/68 (ATCC VR-1679). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-5109