Learn More
Medchemexpress LLC Influenza A virus-IN-1 | 2250313-14-3 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13117910MM/1ML
Encompass_Preferred
Influenza A virus-IN-1 is a dihydropyrrolidones derivative and a potent inhibitor against wide subtypes of influenza A virus (IAV) with IC50 values from 3.11 μM to 7.13 μM. It efficiently inhibits replication of IAV and up-regulates the expression of key antiviral cytokines IFN-β and antiviral protein MxA.
  • Dihydropyrrolidones derivative
  • Potent inhibitor against wide subtypes of influenza A virus (IAV)
  • Efficiently inhibits replication of IAV
  • Up-regulates expression of key antiviral cytokines IFN-β
  • Up-regulates antiviral protein MxA
Catalog No. 50-003-41143