Learn More
ATCC Influenza B virus; B/Hong Kong/5/72
Supplier:  ATCC VR823
Encompass
Influenza B virus strain B/Hong Kong/5/72 is propagated in SPF chicken embryonated eggs. This whole-genome sequenced strain was deposited by the Centers for Disease Control and Prevention and has applications in respiratory and infectious disease research.
Catalog No. 50-238-4048
 Restricted Item