Learn More
Andwin Scientific INFRARED HEAT LAMP
SDP
Supplier:  Andwin Scientific 5930C40
Encompass_Preferred
Uses 250 watt infrared lamp bulb Converging infrared rays prevent the creeping of salts on the walls of vessels Recommended for evaporation, drying precipitates, moisture determinations and several other similar processes Vented aluminum shade has removable wire guard Shade is attached by a swivel joint to extendible, two-segment support arm with spring-balanced hinges Support arm is swivel-mounted on an adjustable clamp for attaching lamp to shelf or table Glides easily to desired elevation and angle
Catalog No. NC9917836