Learn More
TARGETMOL CHEMICALS INC INGENOL 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5S201710MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Ingenol ((-)-Ingenol) mebutate is an effective treatment for actinic keratosis. 2. Formulating novel derivatives from Ingenol esters may be an innovative approach to develop new lead compounds to reactivate latent HIV. 3. Ingenol is a weak PKC activator and possesses important anticancer activity. purity: 99%
Catalog No. 50-257-6633