Learn More
Selleck Chemical LLC Ingliforib S0031-25MG
SDP
Supplier:  Selleck Chemical LLC S003125MG
Encompass_Preferred
Ingliforib (CP 368296) is a glycogen phosphorylase (GP) inhibitor with IC50s of 52 352 and 150 nM for liver muscle and brain glycogen phosphorylase and has cardioprotective activity
Catalog No. 50-000-40224