Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Inhibin beta A Antibody (YA2780) | 100 UL
SDP

Supplier:  Medchemexpress LLC HYP83035100UL

Encompass_Preferred

The Inhibin beta A Antibody (YA2780) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Inhibin beta A. This antibody is developed against a protein from the TGF-beta superfamily, which plays a role in activating and inhibiting follicle stimulating hormone secretion, as well as eye, tooth, and testis development.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Application: Western Blot (WB)
  • Clonality: Monoclonal
  • Immunogen: Inhibin beta A
  • Isotype: IgG
  • Observed molecular weight: 43 kDa
  • Purity: Affinity purified
  • Storage: -20°C for 1 year, avoiding repeated freeze/thaw cycles

Catalog No. 50-004-44351


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.