Learn More
Apexbio Technology LLC Protease and Phosphatase Inhibitor Cocktail (EDTA Free, 100X in ddH2O)(Synonyms: Protease and Phosphatase Inhibitor Mix, EDTA-Free Protease and Phosphatase Inhibitor Cocktail), 50X1ml.
SDP
Supplier:  Apexbio Technology LLC K400650
Encompass_Preferred
Protease and Phosphatase Inhibitor Cocktail (EDTA Free 100X in ddH2O) is an aqueous concentrated mixture designed for inhibition of protease and phosphatase activities during protein extraction procedures The included protease inhibitors target aminopeptidases as well as cysteine and serine proteases while the phosphatase inhibitors suppress activity of serine/threonine and protein tyrosine phosphatases It is intended for protecting protein integrity and phosphorylation status in lysates derived from primary cells cultured mammalian cells animal or plant tissues yeast and bacteria The formulation is free of EDTA to avoid interference with metal-dependent assays with an option available containing EDTA separately (K4005)
Catalog No. 50-000-61396