Learn More
STEMCELL Technologies Latrunculin A, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 1000563
Encompass
Latrunculin A is a 2-thiazolidinone macrolide isolated from the Red Sea sponge Latrunculia magnifica (Coué et al.). It forms a 1:1 molar complex with monomeric G-actin and inhibits actin polymerization in vitro (Kd = 0.2 μM; Coué et al.). Actins are important for numerous cellular functions such as endocytosis, cell migration, and tumor invasion. This product is supplied as a 100 μg/mL solution in ethanol.CANCER RESEARCH· Disrupts actin cytoskeleton in tumor cells (Hayot et al.).
Catalog No. 50-206-9371