Learn More
STEMCELL Technologies Z-VAD-FMK, Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 1000535
Encompass
Z-VAD-FMK is a cell-permeable synthetic peptide that inhibits caspases and blocks caspase-mediated apoptosis in vivo (Garcia-Calvo et al.; Xiang et al.). Z-VAD-FMK prevents differentiation and enhances the freeze-thaw survival rate of human embryonic stem cells when subjected to cryopreservation conditions (Heng et al.).CANCER RESEARCH· Blocks Fas-induced apoptosis in T lymphocytes (Chow et al.).
Catalog No. 50-206-9345