Learn More
Apexbio Technology LLC Iniparib (BSI-201) 160003-66-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A41575.1
Encompass_Preferred
Iniparib (BSI-201) (CAS 160003-66-7) is a small-molecule inhibitor initially characterized as targeting poly(ADP-ribose) polymerase-1 (PARP1) It is designed to inhibit PARP1 by promoting oxidation of zinc finger motifs leading to zinc ejection and suppression of enzyme activity Iniparib exerts its biological activity primarily through metabolism into a C-nitroso intermediate which modifies cysteine residues in multiple proteins In cellular assays Iniparib demonstrates growth-inhibitory activity with reported IC50 values approximately ranging from 40 to 100 M in cancer cell lines Based on these pharmacological properties Iniparib holds research potential in oncology studies particularly in treatment-resistant and solid tumor models
Catalog No. 50-000-94011