Learn More
Medchemexpress LLC Iniparib (BSI-201) | 160003-66-7 | 99.7% | 200 MG
SDP
Supplier:  Medchemexpress LLC HY12015200MG
Encompass_Preferred
Iniparib (BSI-201) is an irreversible poly(ADP-ribose) polymerase 1 (PARP1) inhibitor supplied for research use. It is used in preclinical and translational studies, notably in triple-negative breast cancer research, and is provided as a purified solid suitable for laboratory experiments.
  • Irreversible inhibitor of PARP1.
  • Commonly used in triple-negative breast cancer research.
  • Supplied as a high-purity research-grade compound.
Catalog No. 50-002-33413