Learn More
Medchemexpress LLC Inixaciclib | 2370913-42-9 | 99.9% | C26H30F2N6O | 100 MG

Supplier: Medchemexpress LLC HY45661100MG
Inixaciclib is a potent CDK inhibitor, which can be used for anticancer research.
- Potent CDK inhibitor
- Used for anticancer research
- White to off-white solid
- Molecular weight: 480.55
- Purity: 99.9%
- Powder storage: -20°C for 3 years, 4°C for 2 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.